Tisk…

1,2,4-Trimethylcyclohexane (mixture of stereoisomers), Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL677345-100MLEA 5670 CZK
SIAL677345-100ML
1,2,4-Trimethylcyclohexane (mixture of stereoisomers), Sigma-Aldrich®
1,2,4-Trimethylcyclohexane (mixture of stereoisomers)
Vzorec: C₉H₁₈
MW: 126,24 g/mol
Bod varu: 141 °C (1013 hPa)
Bod tání: –86,4 °C
Hustota: 0,7652 g/cm³ (20 °C)
MDL Number: MFCD00019385
CAS číslo: 2234-75-5
EINECS: 218-783-4
UN: 3295
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR