Tisk…

3,5-Dimethyl-2-cyclohexen-1-one, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIALD154008-5GEA 0 CZK
SIALD154008-5G
3,5-Dimethyl-2-cyclohexen-1-one, Sigma-Aldrich®
3,5-Dimethyl-2-cyclohexen-1-one
Vzorec: C₈H₁₂O
MW: 124,18 g/mol
Bod varu: 211 °C (1013 hPa)
Hustota: 0,88 g/cm³ (20 °C)
MDL Number: MFCD00001583
CAS číslo: 1123-09-7
EINECS: 214-369-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR