Tisk…

5,5-Dimethylbarbituric acid, Sigma-Aldrich®

Dodavatel: Merck

SIAL39566-10GEA 1700 CZK
SIAL39566-10G
5,5-Dimethylbarbituric acid, Sigma-Aldrich®
5,5-Dimethylbarbituric acid
Vzorec: C₆H₈N₂O₃
MW: 156,14 g/mol
Storage Temperature: Ambient
CAS číslo: 24448-94-0
EINECS: 246-265-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR