Tisk…

2-Amino-9-fluorenone, Sigma-Aldrich®


M-Clarity™ quality level = MQ100

SIALA55802-10GEA 12240 CZK
SIALA55802-10G SIALA55802-1G
2-Amino-9-fluorenone, Sigma-Aldrich®
2-Amino-9-fluorenone
Vzorec: C₁₃H₉NO
MW: 195,22 g/mol
Bod varu: 361 °C (1013 hPa)
Bod tání: 163 °C
MDL Number: MFCD00001157
CAS číslo: 3096-57-9
EINECS: 221-445-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR