Tisk…

2-Methoxyfuran, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL138274-5GEA 5980 CZK
SIAL138274-5G
2-Methoxyfuran, Sigma-Aldrich®
2-Methoxyfuran
Vzorec: C₅H₆O₂
MW: 98,1 g/mol
Bod varu: 92…110 °C (1013 hPa)
Hustota: 1,0646 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00003227
CAS číslo: 25414-22-6
EINECS: 246-953-8
UN: 3271
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR