Tisk…

Sodium hexafluoroaluminate, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ100

SIAL230049-25GEA 6580 CZK
SIAL230049-25G
Sodium hexafluoroaluminate, Sigma-Aldrich®
Sodium hexafluoroaluminate
Vzorec: AlF₆Na₃
MW: 209,94 g/mol
Bod tání: 977 °C
Hustota: 2,95 g/cm³ (20 °C)
MDL Number: MFCD00003507
CAS číslo: 13775-53-6
EINECS: 237-410-6
UN: 3260
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR