Tisk…

Rubidium perchlorate, anhydrous ≥99.5% (metals basis)

Dodavatel: Thermo Scientific
Nebezpečí

11628.09EA 2090 CZK
11628.09
Rubidium perchlorate, anhydrous ≥99.5% (metals basis)
Rubidium perchlorate
Vzorec: ClO₄Rb
MW: 184,92 g/mol
Bod tání: 281 °C
Hustota: 2,878 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016294
CAS číslo: 13510-42-4
EINECS: 236-840-1
UN: 1481
ADR: 5.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR