Tisk…

Disodium fluorophosphate, Sigma-Aldrich®

Dodavatel: Merck
Varování


M-Clarity™ quality level = MQ100

SIAL344443-500GEA 1660 CZK
SIAL344443-500G
Disodium fluorophosphate, Sigma-Aldrich®
Disodium fluorophosphate
Vzorec: FNa₂O₃P
MW: 143,95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00014248
CAS číslo: 10163-15-2
EINECS: 233-433-0
UN: 3288
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR