Tisk…
Dihydrogenfosforečnan draselný, ULTREX®, Ultrapure, J.T.Baker®
Dodavatel: Avantor
Varování
4921-04EA 12850 CZK
4921-04 4921-07
Dihydrogenfosforečnan draselný, ULTREX®, Ultrapure, J.T.Baker®
Dihydrogenfosforečnan draselný
Vzorec: KH₂PO₄
MW: 136,09 g/mol
Bod varu: ~100 °C (1013 hPa)
Bod tání: 253 °C
Hustota: 2,34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011401
CAS číslo: 7778-77-0
EINECS: 231-913-4
REACH: 01-2119490224-41
Merck Index: 12,07829
Objednat
Výsledky specifikačního testu
For Laboratory,Research,or Manufacturing Use
Assay (KH₂PO₄) (dried basis), For Information Only
Loss on Drying at 105°C, For Information Only
Particulate Matter, For Information Only
pH of 5% Solution at 25°C, For Information Only
Non-Metallic Impurities - Arsenic (As), For Information Only
Non-Metallic Impurities - Fluoride (F), For Information Only
Non-Metallic Impurities - Halide (as Cl), For Information Only
Non-Metallic Impurities - Nitrogen Compounds (as N), For Information Only
Non-Metallic Impurities - Silicon (Si), For Information Only
Non-Metallic Impurities - Sulfate (SO₄), For Information Only
Metallic Impurities - Aluminum (Al), For Information Only
Metallic Impurities - Barium (Ba), For Information Only
Metallic Impurities - Bismuth (Bi), For Information Only
Metallic Impurities - Cadmium (Cd), For Information Only
Metallic Impurities - Calcium (Ca), For Information Only
Metallic Impurities - Chromium (Cr), For Information Only
Metallic Impurities - Cobalt (Co), For Information Only
Metallic Impurities - Copper (Cu), For Information Only
Metallic Impurities - Iron (Fe), For Information Only
Metallic Impurities - Lead (Pb), For Information Only
Metallic Impurities - Magnesium (Mg), For Information Only
Metallic Impurities - Manganese (Mn), For Information Only
Metallic Impurities - Mercury (Hg), For Information Only
Metallic Impurities - Molybdenum (Mo), For Information Only
Metallic Impurities - Nickel (Ni), For Information Only
Metallic Impurities - Niobium (Nb), For Information Only
Metallic Impurities - Silver (Ag), For Information Only
Metallic Impurities - Sodium (Na), For Information Only
Metallic Impurities - Strontium (Sr), For Information Only
Metallic Impurities - Tin (Sn), For Information Only
Metallic Impurities - Titanium (Ti), For Information Only
Metallic Impurities - Vanadium (V), For Information Only
Metallic Impurities - Zinc (Zn), For Information Only
Metallic Impurities - Zirconium (Zr), For Information Only
UV Absorbance (1 M Aqueous Solution) (1.00-cm cell vs. water) - 400 nm, For Information Only
UV Absorbance (1 M Aqueous Solution) (1.00-cm cell vs. water) - 280 nm, For Information Only
UV Absorbance (1 M Aqueous Solution) (1.00-cm cell vs. water) - 254 nm, For Information Only
UV Absorbance (1 M Aqueous Solution) (1.00-cm cell vs. water) - 220 nm, For Information Only
UV Absorbance (1 M Aqueous Solution) (1.00-cm cell vs. water) - 215 nm, For Information Only
Certificate Provided Reports Actual Lot Analysis