Potassium hexaiodoplatinate(IV) (≥18.2% Pt)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16905-14-9
:
Formula:I₆K₂Pt
MDL Number:MFCD00049660
:1034.7 g/mol
EINECS:240-954-7
:GHS07
Storage Temperature:Ambient
Related Information
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: 12642.03Local Article Number: ALFA12642.03
- Supplier Article Number: 12642.06Local Article Number: ALFA12642.06