Oxid hlinitý substrate 12.7x12.7x1 mm, polished one side M plane
Synonyms:
Alumina C
43246.KTEA
5930
CZK
43246.KT
Oxid hlinitý substrate 12.7x12.7x1 mm, polished one side M plane
Oxid hlinitý
Vzorec:
Al₂O₃ MW: 101,96 g/mol Bod varu: 2980 °C (1013 hPa) Bod tání: 2045 °C Hustota: 3,94 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003424 CAS číslo: 1344-28-1 EINECS: 215-691-6 REACH: 01-2119529248-35 Merck Index: 13,00355 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...