Tisk…

Cyclopentadienylthallium, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL155349-10GEA 6170 CZK
SIAL155349-10G
Cyclopentadienylthallium, Sigma-Aldrich®
Cyclopentadienylthallium
Vzorec: C₅H₅Tl
MW: 269,48 g/mol
Bod varu: 90…110 °C (1 mmHg)
Bod tání: 75 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00134141
CAS číslo: 34822-90-7
EINECS: 252-229-2
UN: 1707
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR