Tisk…

Silicon(II) oxide ≥99.99% (metals basis)

Dodavatel: Thermo Scientific
Varování

36347.22EA 16240 CZK
36347.22 36347.14
Silicon(II) oxide ≥99.99% (metals basis)
Silicon(II) oxide
Vzorec: SiO
MW: 44,08 g/mol
Bod varu: 1880 °C
Bod tání: >1702 °C
Hustota: 2,13
Storage Temperature: Ambient
MDL Number: MFCD00151536
CAS číslo: 10097-28-6
EINECS: 233-232-8
Merck Index: 12,08640

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR