Tisk…

(3-Bromopropyl)trimethoxysilane, Sigma-Aldrich®

Dodavatel: Merck

SIAL18265-5MLEA 6730 CZK
SIAL18265-5ML
(3-Bromopropyl)trimethoxysilane, Sigma-Aldrich®
(3-Bromopropyl)trimethoxysilane
Vzorec: BrH₂CCH₂CH₂Si(OCH₃)₃
MW: 243,17 g/mol
MDL Number: MFCD00053894
CAS číslo: 51826-90-5
EINECS: 257-453-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR