Tisk…

(3-Glycidoxypropyl)triethoxysilane, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL50059-50MLEA 16630 CZK
SIAL50059-50ML
(3-Glycidoxypropyl)triethoxysilane, Sigma-Aldrich®
(3-Glycidoxypropyl)triethoxysilane
Vzorec: C₁₂H₂₆O₅Si
MW: 278,42 g/mol
MDL Number: MFCD01311705
CAS číslo: 2602-34-8
EINECS: 220-011-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR