Tisk…

Octyltrimethoxysilane, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL376221-25MLEA 6090 CZK
SIAL376221-25ML
Octyltrimethoxysilane, Sigma-Aldrich®
Octyltrimethoxysilane
Vzorec: C₁₁H₂₆O₃Si
MW: 234,41 g/mol
MDL Number: MFCD00043053
CAS číslo: 3069-40-7
EINECS: 221-338-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR