Tisk…

Triisobutylsilane, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL278785-10GEA 6360 CZK
SIAL278785-10G SIAL278785-50G
Triisobutylsilane, Sigma-Aldrich®
Triisobutylsilane
Vzorec: C₁₂H₂₈Si
MW: 200,44 g/mol
MDL Number: MFCD00010340
CAS číslo: 6485-81-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR