Tisk…

Phenyltin trichloride

Dodavatel: Thermo Scientific
Nebezpečí

71157.14EA 9920 CZK
71157.14 71157.06
Phenyltin trichloride
Phenyltin trichloride
Vzorec: C₆H₅Cl₃Sn
MW: 302,17 g/mol
Bod varu: 109…110 °C (10 mmHg)
Hustota: 1,84
Storage Temperature: Ambient
MDL Number: MFCD00000513
CAS číslo: 1124-19-2
EINECS: 214-393-3
UN: 2788
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR