Tisk…
Pipette tips, epT.I.P.S. Biopur® (General Lab Product)
Dodavatel: Eppendorf
epT.I.P.S. Biopur®
EPPE0030010.027EA 958 CZK
EPPE0030010.027 EPPE0030010.060 613-6818 EPPE0030010.043
Pipette tips, epT.I.P.S. Biopur® (General Lab Product)
Špičky pipetovací
These sterile tips are free of DNA, DNase, RNase and pyrogens. Conform to the highest standards for use in medical and pharmaceutical environments, the food industry, and in molecular and cellular biology applications.
epT.I.P.S., in combination with adequate pipettes, are intended for dispensing liquids in the volume range of 0.1 µl – 10 ml. In vivo applications (applications in or on the human body) are not allowed. epT.I.P.S. and the appropriate pipettes only must be used by correspondingly trained specialists. The user must have read thoroughly the operating instructions of the pipette and the instructions for use of the epT.I.P.S. and must have made himself familiar with the functioning of the pipette with the pipette tips.
Certifikace: Without CE marking.
Balení: Available as epT.I.P.S.® Singles (packaged individually) or as epT.I.P.S.® Racks (boxes containing racks of 96, 48 or 24 tips depending on tip volume).