Tisk…

Phosphorus(III) iodide, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL241555-10GEA 2800 CZK
SIAL241555-10G
Phosphorus(III) iodide, Sigma-Aldrich®
Phosphorus(III) iodide
Vzorec: I₃P
MW: 411,69 g/mol
Bod tání: 60 °C
Hustota: 4,18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011442
CAS číslo: 13455-01-1
EINECS: 236-647-2
UN: 3260
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR