Tisk…

(Isopentyl)triphenylphosphonium bromide, Sigma-Aldrich®

Dodavatel: Merck
Varování


M-Clarity™ quality level = MQ100

SIAL347779-25GEA 1130 CZK
SIAL347779-25G
(Isopentyl)triphenylphosphonium bromide, Sigma-Aldrich®
(Isopentyl)triphenylphosphonium bromide
Vzorec: (H₃C)₂HCCH₂CH₂P(C₆H₅)₃Br
MW: 413,34 g/mol
MDL Number: MFCD00031594
CAS číslo: 28322-40-9
EINECS: 248-966-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR