Tisk…

Acetylthiocholine chloride

MOLE61538581-1GEA 5550 CZK
MOLE61538581-1G
Acetylthiocholine chloride
Acetylthiocholine chloride
Vzorec: H₃CC(S)O(CH₂)₂N(CH₃)₃Cl
MW: 197,73 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00038727
CAS číslo: 6050-81-3
EINECS: 227-953-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR