Tisk…

Tetrabutylammonium fluoride trihydrate

MOLE26809718-100GEA 4200 CZK
MOLE26809718-100G
Tetrabutylammonium fluoride trihydrate
Tetrabutylammonium fluoride trihydrate
Vzorec: C₁₆H₃₆FN·3H₂O
MW: 315,51 g/mol
Bod tání: 62…63 °C
MDL Number: MFCD00149981
CAS číslo: 87749-50-6
EINECS: 207-057-2
UN: 2811
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR