Tisk…

Fenylhydrazin analytical reagent, Supelco®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ100

1.07251.0100EA 2550 CZK
1.07251.0100
Fenylhydrazin analytical reagent, Supelco®
Fenylhydrazin
Vzorec: C₆H₈N₂
MW: 108,14 g/mol
Bod varu: 243 °C (1013 hPa)
Bod tání: 19,6 °C
Hustota: 1,0978 g/cm³ (20 °C)
Bod vzplanutí: 86 °C
MDL Number: MFCD00007573
CAS číslo: 100-63-0
EINECS: 202-873-5
UN: 2572
ADR: 6.1,II
Merck Index: 12,07447

Objednat

Výsledky specifikačního testu

Hill Formula C₆H₈N₂
Chemical formula C₆H₅NHNH₂
Molar Mass 108.14 g/mol
Boiling point 244 °C (1013 hPa)
Density 1.10 g/cm³ (20 °C)
Explosion limit 1.1 %(V)
Flash point 88 °C
Ignition temperature 195 °C
Melting Point 19.6 °C
pH value (H₂O, 25 °C) alkaline,(undiluted)
Vapor pressure 0.1 hPa (20 °C)
Solubility 145 g/l (25 °C)
LD 50 oral LD50 Rat 188 mg/kg
Storage class 6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK WGK 3 highly water endangering

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR