Tisk…

2-Ethylimidazole, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

SIAL239348-500GEA 2430 CZK
SIAL239348-500G SIAL239348-100G
2-Ethylimidazole, Sigma-Aldrich®
2-Ethylimidazole
Vzorec: C₅H₈N₂
MW: 96,13 g/mol
Bod varu: 268 °C (1013 hPa)
Hustota: 1,03 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005192
CAS číslo: 1072-62-4
EINECS: 214-011-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR