Tisk…

Ytterbium(III) chloride, Sigma-Aldrich®

Dodavatel: Merck
Varování


M-Clarity™ quality level = MQ100

SIAL439614-5GEA 10010 CZK
SIAL439614-5G
Ytterbium(III) chloride, Sigma-Aldrich®
Ytterbium(III) chloride
Vzorec: Cl₃Yb
MW: 279,4 g/mol
Bod tání: 860 °C
MDL Number: MFCD00011288
CAS číslo: 10361-91-8
EINECS: 233-800-5
Merck Index: 13,10014

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR