Tisk…

Terbium(III,IV) oxide, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL204579-2GEA 6800 CZK
SIAL204579-2G
Terbium(III,IV) oxide, Sigma-Aldrich®
Terbium(III,IV) oxide
Vzorec: O₇Tb₄
MW: 747,7 g/mol
Bod tání: 2283…2323 °C
MDL Number: MFCD00083151
CAS číslo: 12037-01-3
EINECS: 234-856-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR