Tisk…

2-Naphthyl isocyanate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL472514-1GEA 4360 CZK
SIAL472514-1G
2-Naphthyl isocyanate, Sigma-Aldrich®
2-Naphthyl isocyanate
Vzorec: C₁₁H₇NO
MW: 169,18 g/mol
Bod varu: 125…127 °C (3 mmHg)
Bod tání: 57 °C
MDL Number: MFCD00014319
CAS číslo: 2243-54-1
EINECS: 218-815-7
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR