Tisk…

Sodium bis(trimethylsilyl)amide 95+%, pure

Dodavatel: Thermo Scientific
Nebezpečí

ACRO215961000EA 13500 CZK
ACRO215961000 ACRO215960050 ACRO215960250
Sodium bis(trimethylsilyl)amide 95+%, pure
Sodium bis(trimethylsilyl)amide
Vzorec: C₆H₁₈NNaSi₂
MW: 183,38 g/mol
Bod tání: 171…175 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009835
CAS číslo: 1070-89-9
EINECS: 213-983-8
UN: 3096
ADR: 8,I

Objednat

Výsledky specifikačního testu

Appearance White to light yellow to light beige Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 95.0 to 105.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR