Tisk…

Sodium bis(trimethylsilyl)amide ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

33505.09EA 2910 CZK
33505.09 33505.04
Sodium bis(trimethylsilyl)amide ≥98%
Sodium bis(trimethylsilyl)amide
Vzorec: C₆H₁₈NNaSi₂
MW: 183,38 g/mol
Bod tání: 171…175 °C
Hustota: 1,065 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009835
CAS číslo: 1070-89-9
EINECS: 213-983-8
UN: 3096
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR