Tisk…
Microinjectors, CellTram® 4m
Dodavatel: Eppendorf
CellTram®
EPPE5196000.048EA 0 CZK
EPPE5196000.048 EPPE5196000.021
Microinjectors, CellTram® 4m
Mikroinjektory Mikroinjektory manuální
The CellTram® 4 m Air and CellTram® 4 m Oil are manual microinjectors for use in assisted reproductive technology. The CellTram® 4 m was designed and manufactured for injection and aspiration of cells or cellular parts in the context of ex-vivo micromanipulations in combination with suitable microcapillaries in the area of human reproductive medicine.
  • Non-hydraulic system for oil-free manipulation and injection of cells (CellTram® 4 m Air)
  • Piston position scale enables setting of air volume and injection characteristics according to individual needs (CellTram® 4r Air)
  • Improved oil-filling system for minimal interruption of the workflow (CellTram® 4 m Oil)
  • Coarse and fine drive for precise and intuitive sample control and improved rotary knob design for better handling and easier cleaning
  • Capillary holder 4 with markings for easier positioning and grip head 4 system for simple capillary change and higher user safety
  • Position indicators and marking for in/out to reduce risk of user errors
  • Stable, non slip base plate
The CellTram® 4 m Air is a pneumatic injector for oil-free micromanipulation and gentle holding of cells in suspension. Due to its excellent performance, it is also highly suitable for uptake and injection of cells (e.g. sperm). For this application, its performance is comparable to that of oil-filled injectors.
The CellTram® 4 m Oil is a hydraulic, oil-filled microinjector optimised for precise pressure control, leakage-free work and easy oil filling.
Both systems offer highly reliable performance with minimal maintenance.