Tisk…

(R,R)-(+)-Bis(ɑ-methylbenzyl)amine hydrochloride 97% (by GC), Sigma-Aldrich®

Dodavatel: Merck
Varování


M-Clarity™ quality level = MQ100

SIAL461520-5GEA 7040 CZK
SIAL461520-5G
(R,R)-(+)-Bis(ɑ-methylbenzyl)amine hydrochloride 97% (by GC), Sigma-Aldrich®
(R,R)-(+)-Bis(alpha-methylbenzyl)amine hydrochloride
Vzorec: C₁₆H₁₉N
MW: 261,79 g/mol
Bod tání: 260…263 °C (lit.)
MDL Number: MFCD00216672
CAS číslo: 82398-30-9

Objednat

Výsledky specifikačního testu

Infrared Spectrum Conforms to Structure
Optical Rotation (c = 3%, Ethanol ) 71.0 - 75.0°
Titration by AgNO₃ 96.5 - 103.5%
Purity (GC) ≥96.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR