Tisk…
Toggle anchor, DBI-SALA®, Saflok®, 3M™
CPSD2100103EA 0 CZK
CPSD2100103
Toggle anchor, DBI-SALA®, Saflok®, 3M™
DBI-SALA® Saflok® Toggle anchor for hollow concrete and steel, fits into a 19 mm hole, with swivelling D-ring.
  • Use vertically in wall or overhead ceiling applications
  • Swivelling D-ring tie-off point
  • Activator cable for toggle mode selection
  • Toggle position indicator
  • Portable and reusable
This anchorage connector is made from top quality materials guaranteeing high strength and durability to stand up to the toughest environments.
Certifikace: EN 795, Class B