Labels for BradyJet J2000 Lab printer
Supplier: Brady
Certificates
About this item
These PP BradyJet J2000 Lab printer labels come in matching sizes and shapes for most sample containers including tubes, vials, cryo containers, dishes, plates and general glassware.
- Designed to stay attached and remain legible
- Resist liquid nitrogen, freezers, hot water baths and autoclaves
They can also resist common laboratory chemicals like acetone, ethanol, toluene, xylene and IPA.
Related Information
Product Information
- Catalog #Supplier #TypeModelMaterialPrinting methodColourAvailabilityPrice
- Printer labelsPrinter labelsBRDY150005150005B-7425JJ20-152-7425JPPWhite1 * 1 Roll5 700,00 KčSupplier Article Number: 150005Local Article Number: BRDY150005
Specifications
Environmentally Preferable - Printer labelsPrinter labelsBRDY150007150007B-7425JJ20-257-7425JPPWhite1 * 1 Roll6 800,00 KčSupplier Article Number: 150007Local Article Number: BRDY150007
Specifications
Environmentally Preferable - Labels with 9 mm dotLabels with 9 mm dotBRDY150023150023B-7425JJ20-249-7425JPPWhite1 * 1 Roll6 840,00 KčSupplier Article Number: 150023Local Article Number: BRDY150023
Specifications
Environmentally Preferable - Printer labelsPrinter labelsBRDY150006150006B-7425JJ20-59-7425JPPWhite1 * 1 Roll6 210,00 KčSupplier Article Number: 150006Local Article Number: BRDY150006
Specifications
Environmentally Preferable - Labels with 11 mm dotLabels with 11 mm dotBRDY150033150033B-7425JJ20-251-7425JPPWhite1 * 1 Roll6 730,00 KčSupplier Article Number: 150033Local Article Number: BRDY150033
Specifications
Environmentally Preferable - Printer labelsPrinter labelsBRDY150015150015B-7425JJ20-88-7425JPPWhite1 * 1 Roll12 110,00 KčSupplier Article Number: 150015Local Article Number: BRDY150015
Specifications
Environmentally Preferable - Printer labelsPrinter labelsBRDY150025150025B-7425JJ20-179-7425JPPWhite1 * 1 Roll6 110,00 KčSupplier Article Number: 150025Local Article Number: BRDY150025
Specifications
Environmentally Preferable - Label printer tapeLabel printer tapeBRDY150542150542B-7425JJ20C-1125-7425JPPWhite1 * 1 KS6 470,00 KčSupplier Article Number: 150542Local Article Number: BRDY150542
Specifications
Environmentally Preferable - Printer labelsPrinter labelsBRDY150016150016B-7425JJ20-256-7425JPPWhite1 * 1 Roll5 570,00 KčSupplier Article Number: 150016Local Article Number: BRDY150016
Specifications
Environmentally Preferable - Printer labelsPrinter labelsBRDY150020150020B-7425JJ20-37-7425JPPWhite1 * 1 Roll5 940,00 KčSupplier Article Number: 150020Local Article Number: BRDY150020
Specifications
Environmentally Preferable - Printer labelsPrinter labelsBRDY150019150019B-7425JJ20-136-7425JPPWhite1 * 1 Roll8 620,00 KčSupplier Article Number: 150019Local Article Number: BRDY150019
Specifications
Environmentally Preferable - Printer labelsPrinter labelsBRDY150022150022B-7425JJ20-235-7425JPPWhite1 * 1 Roll3 690,00 KčSupplier Article Number: 150022Local Article Number: BRDY150022
Specifications
Environmentally Preferable - Printer labelsPrinter labelsBRDY150028150028B-7425JJ20-258-7425JPPWhite1 * 1 Roll5 870,00 KčSupplier Article Number: 150028Local Article Number: BRDY150028
Specifications
Environmentally Preferable - Printer labelsPrinter labelsBRDY150011150011B-7425JJ20-151-7425JPPWhite1 * 1 Roll5 640,00 KčSupplier Article Number: 150011Local Article Number: BRDY150011
Specifications
Environmentally Preferable - Printer labelsPrinter labelsBRDY150024150024B-7425JJ20-53-7425JPPWhite1 * 1 Roll8 140,00 KčSupplier Article Number: 150024Local Article Number: BRDY150024
Specifications
Environmentally Preferable - Printer labelsPrinter labelsBRDY150018150018B-7425JJ20-137-7425JPPWhite1 * 1 Roll4 650,00 KčSupplier Article Number: 150018Local Article Number: BRDY150018
Specifications
Environmentally Preferable - Printer labelsPrinter labelsBRDY150543150543B-7425JJ20-260-7425JPPWhite1 * 340 KS9 690,00 KčSupplier Article Number: 150543Local Article Number: BRDY150543
Specifications
Environmentally Preferable - Label printer tapeLabel printer tapeBRDY150547150547B-7425JJ20C-2250-7425JPPWhite1 * 1 KS9 570,00 KčSupplier Article Number: 150547Local Article Number: BRDY150547
Specifications
Environmentally Preferable - Printer labelsPrinter labelsBRDY150541150541B-7425JJ20-261-7425JPPWhite1 * 270 KS10 680,00 KčSupplier Article Number: 150541Local Article Number: BRDY150541
Specifications
Environmentally Preferable - Printer labelsPrinter labelsBRDY150544150544B-7425JJ20-262-7425JPPWhite1 * 200 KS14 760,00 KčSupplier Article Number: 150544Local Article Number: BRDY150544
Specifications
Environmentally Preferable - Printer labelsPrinter labelsBRDY150545150545B-7425JJ20-18-7425JPPWhite1 * 1 Roll12 520,00 KčSupplier Article Number: 150545Local Article Number: BRDY150545
Specifications
Environmentally Preferable
Ink pads
Product Information
- Catalog #Supplier #TypeModelMaterialPrinting methodColourAvailabilityPrice
- Ink padsInk padsBRDY150163150163J20-PADWhite1 * 3 KS607,00 KčSupplier Article Number: 150163Local Article Number: BRDY150163