Tisk…
Temperature and pressure logger set, EBI 12
Dodavatel: Ebro
EBRO1250-1111EA 177030 CZK
EBRO1250-1111
Temperature and pressure logger set, EBI 12
These logger set is specially designed for temperature and pressure monitoring and as well as for A₀ value calculation in steam sterilisers, washer-disinfectors and washer-disinfectors for endoscopes.
Informace o dodávce: The set includes with EBI 12-TP231 pressure/temperature data logger with Luer-Lock connection, AL 101 silicone protection box for loggers, Winlog.med software, EBI IF 150 interfaces, carrying case 'SYSTAINER' and pH and conductivity tester.