The frameiof the of the βγoxene® goggle model 611is made from a special material which is resistant to several sterlisation cycles, while the innovative βyoxene lens allows to keep the original transparency also after 20 cycles of sterilisation according to the common techniques β or γ rays. βγoxene® looses only 1,4% of the light transmittance, a percentage which is lost from the polycarbonate only after one single cycle of sterilisation.
- βγoxene® lenses
- Available with directly or indirectly ventilation
- Elastic band in anti-static silicone
- RX insert
Designed to be used in rooms with contamination control (sterile and cleanroom) and in aseptic environments.