Tisk…
Orange filter for myECL™ imager
Dodavatel: Thermo Fisher Scientific
Pierce
PIER62238EA 28920 CZK
PIER62238
Orange filter for myECL™ imager
Systémy fotodokumentační
Thermo Scientific™ 565 to 620 nm bandpass filter is a threaded, 50 mm diameter glass filter. This product is a replacement for the pre-installed orange filter in the myECL™ Imager, and is used for detection of colorimetric protein stains (silver, coomassie) and wavelengths of common fluorescent nucleic acid gel stains (e.g., ethidium bromide) and protein stains, such as SYPRO® Orange Protein Gel Stain.