General purpose gloves, TEGERA® 888
Supplier: EJENDALS
Certificates
About this item
These unlined gloves are made of a combination of full grain cowhide leather (50%), cotton (49%) and natural latex (1%).
- Reinforced index finger, reinforced fingers and thumb
- Elasticated 360° fastening
- High level of protection with good fingertip sensitivity
- Durable and good fit
- Ideal for works with medium weight
The gloves prevent risks of abrasion injuries, blisters, grazes, scratches, lacerations and contact with dirt.
Primary areas of use: Airport work, assembly, cleaning, building and construction, warehouse work, carpentry, installation work, engineering, transport work, machine driving, wood industry work, sanitation and machine operating.
Colour: Grey/blue/black/white
Certifications: EN 388:2016 (2121X); Cat. II.
Specifications
- Catalog No:
- 112-5380
- 112-5384
- 112-5383
- 112-5382
- 112-5381
- 112-5386
- 112-5385
- Model:
- 888
- 888
- 888
- 888
- 888
- 888
- 888
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Material:
- Cotton/leather
- Cotton/leather
- Cotton/leather
- Cotton/leather
- Cotton/leather
- Cotton/leather
- Cotton/leather
- Finish:
- Reinforced fingers
- Reinforced fingers
- Reinforced fingers
- Reinforced fingers
- Reinforced fingers
- Reinforced fingers
- Reinforced fingers
- Cuff style:
- Elastic
- Elastic
- Elastic
- Elastic
- Elastic
- Elastic
- Elastic
- Palm thickness:
- 0,7 - 0,8 mm
- 0,7 - 0,8 mm
- 0,7 - 0,8 mm
- 0,7 - 0,8 mm
- 0,7 - 0,8 mm
- 0,7 - 0,8 mm
- 0,7 - 0,8 mm
- EN norm:
- EN 388:2016 (2121X)
- EN 388:2016 (2121X)
- EN 388:2016 (2121X)
- EN 388:2016 (2121X)
- EN 388:2016 (2121X)
- EN 388:2016 (2121X)
- EN 388:2016 (2121X)
Specifications
Product Information
- Catalog #Supplier #ModelGlove sizeLengthPkAvailabilityPrice
- Supplier Article Number: 888-6Local Article Number: EJEN888-6Specifications:
More
Specifications
Model888Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLatex / latex-freeLatexMaterialCotton/leatherFinishReinforced fingersCuff styleElasticPalm thickness0,7 - 0,8 mmEN normEN 388:2016 (2121X) - Supplier Article Number: 888-7Local Article Number: EJEN888-7Specifications:
More
Specifications
Model888Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLatex / latex-freeLatexMaterialCotton/leatherFinishReinforced fingersCuff styleElasticPalm thickness0,7 - 0,8 mmEN normEN 388:2016 (2121X) - Supplier Article Number: 888-8Local Article Number: EJEN888-8Specifications:
More
Specifications
Model888Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLatex / latex-freeLatexMaterialCotton/leatherFinishReinforced fingersCuff styleElasticPalm thickness0,7 - 0,8 mmEN normEN 388:2016 (2121X) - Supplier Article Number: 888-9Local Article Number: EJEN888-9Specifications:
More
Specifications
Model888Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLatex / latex-freeLatexMaterialCotton/leatherFinishReinforced fingersCuff styleElasticPalm thickness0,7 - 0,8 mmEN normEN 388:2016 (2121X) - Supplier Article Number: 888-10Local Article Number: EJEN888-10Specifications:
More
Specifications
Model888Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLatex / latex-freeLatexMaterialCotton/leatherFinishReinforced fingersCuff styleElasticPalm thickness0,7 - 0,8 mmEN normEN 388:2016 (2121X) - Supplier Article Number: 888-11Local Article Number: EJEN888-11Specifications:
More
Specifications
Model888Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLatex / latex-freeLatexMaterialCotton/leatherFinishReinforced fingersCuff styleElasticPalm thickness0,7 - 0,8 mmEN normEN 388:2016 (2121X) - Supplier Article Number: 888-12Local Article Number: EJEN888-12Specifications:
More
Specifications
Model888Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLatex / latex-freeLatexMaterialCotton/leatherFinishReinforced fingersCuff styleElasticPalm thickness0,7 - 0,8 mmEN normEN 388:2016 (2121X)