Tisk…
Reusable particle filtering respirators, FFP3, Air Plus ProValve
Dodavatel: Moldex
MOLD340801EA 1460 CZK
MOLD340801
Reusable particle filtering respirators, FFP3, Air Plus ProValve
Polomasky a filtrační respirátory Polomasky filtrační opakovatelně použitelné Polomasky
Reusable FFP mask with a very low breathing resistance. Ideal to protect against dust, mist and fume.
  • AirWave® pleated filter for comfortable breathing and fatigue-free work
  • Longer use time, less consumption
  • 3D face seal – like a rubber half mask – improves fit and provides optimum wearing comfort
  • Built-in ProValve, reduces condensation from exhaled air
  • Comfortable full-face seal made of washable TPE material, gentle on the skin
  • ActivForm®: no nose clip needed
Pleated filter technology reduces inhalation resistance by up to 50% whilst maintaining filtration performance. DuraMesh® strong and durable structure, keeps the mask in shape. The full face seal can be wiped clean and gives the option of using the mask for more than one shift.
Length-adjustable loop strap with clip for quick, simple and safe on and off. Masks have passed the Dolomite clogging test, giving the user better breathing resistance for longer.
Certifikace: EN 149:2001+A1:2009