Order Entry
Tube furnaces with stand for horizontal or vertical operation, RT
Tube furnaces with stand for horizontal or vertical operation, RT
Supplier:  NABERTHERM
Certificates
undefined
Tube furnaces with stand for horizontal or vertical operation, RT
Supplier:  NABERTHERM
Tmax 1500 models with B 510 controller
Product Information
  • Catalog #
    Supplier #
    Type
    Max. temp.
    Power
    Ø int. tube
    Tube length
    Heated length
    Availability
    Price
  • 455-2103
    RT512LAN
    RT 50-250/11 B 510 (1 phase)
    1100 °C
    1800 W
    50 mm
    360 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 168 200,00 Kč
    168 200,00 Kč
    1 * 1 KS
       168 200,00 Kč
    Supplier Article Number: RT512LAN
    Local Article Number: NABERT512LAN
    Specifications:
    Specifications
    Environmentally Preferable
  • 455-2098
    RT332LAN
    RT 30-200/15 B 510 (1 phase)
    1500 °C
    1800 W
    30 mm
    360 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 368 880,00 Kč
    368 880,00 Kč
    1 * 1 KS
       368 880,00 Kč
    Supplier Article Number: RT332LAN
    Local Article Number: NABERT332LAN
    Specifications:
    Specifications
    Environmentally Preferable
  • 455-2105
    RT522LAN
    RT 50-250/13 B 510 (1 phase)
    1300 °C
    1800 W
    50 mm
    360 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 223 970,00 Kč
    223 970,00 Kč
    1 * 1 KS
       223 970,00 Kč
    Supplier Article Number: RT522LAN
    Local Article Number: NABERT522LAN
    Specifications:
    Specifications
    Environmentally Preferable
Tmax 1500 models with C 550 controller
Product Information
  • Catalog #
    Supplier #
    Type
    Max. temp.
    Power
    Ø int. tube
    Tube length
    Heated length
    Availability
    Price
  • 455-2099
    RT332OAN
    RT 30-200/15 C 550 (1 phase)
    1500 °C
    1800 W
    30 mm
    360 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 392 470,00 Kč
    392 470,00 Kč
    1 * 1 KS
       392 470,00 Kč
    Supplier Article Number: RT332OAN
    Local Article Number: NABERT332OAN
    Specifications:
    Specifications
    Environmentally Preferable
  • 455-2104
    RT512OAN
    RT 50-250/11 C 550 (1 phase)
    1100 °C
    1800 W
    50 mm
    360 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 191 800,00 Kč
    191 800,00 Kč
    1 * 1 KS
       191 800,00 Kč
    Supplier Article Number: RT512OAN
    Local Article Number: NABERT512OAN
    Specifications:
    Specifications
    Environmentally Preferable
  • 455-2106
    RT522OAN
    RT 50-250/13 C 550 (1 phase)
    1300 °C
    1800 W
    50 mm
    360 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 247 560,00 Kč
    247 560,00 Kč
    1 * 1 KS
       247 560,00 Kč
    Supplier Article Number: RT522OAN
    Local Article Number: NABERT522OAN
    Specifications:
    Specifications
    Environmentally Preferable