Order Entry
VISOCOLOR® Colorimetric Tests, Chlorine
undefined
VISOCOLOR® Colorimetric Tests, Chlorine
Supplier:  MACHEREY-NAGEL
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Model:
    MANA931219
  • Platform:
    Photometric test kits
  • Storage temperature:
    15...25 °C
  • Shelf life (months):
    24
  • Weight:
    145,1 g (Including packaging)
Specifications
Test Kits
Product Information
  • Catalog #
    Supplier #
    Contents
    Range
    Chemical method
    For unit
    Dimensions
    Availability
    Price
  • VISOCOLOR® ECO Chlorine 1, test no. 5-35; Measuring range on PF-12Plus. Range on other photometers can be different
    VISOCOLOR® ECO Chlorine 1, test no. 5-35; Measuring range on PF-12Plus. Range on other photometers can be different
    MANA931035
    931035
    Reagents for 150 tests, colour chart and accessories in a box
    < 0,1 • 0,1 • 0,2 • 0,3 • 0,4 • 0,6 • 0,9 • 1,2 • 2,0 mg/L (Cl₂); 0,05 - 2,00 mg/L (Cl₂); 0,05 - 2,00 mg/L (Cl₂)
    DPD
    PF-12Plus; PF-3 Drinking Water; Advance; PF-3 Pool; Visual
    173×117×42 mm (Including packaging)
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 1 780,00 Kč
    1 780,00 Kč
    1 * 1 KIT
       1 780,00 Kč
    Supplier Article Number: 931035
    Local Article Number: MANA931035
  • VISOCOLOR® ECO free Chlorine 2, test no. 5-16; Measuring range on PF-12Plus. Range on other photometers can be different
    VISOCOLOR® ECO free Chlorine 2, test no. 5-16; Measuring range on PF-12Plus. Range on other photometers can be different
    MANA931016
    931016
    Reagents for 150 tests, colour chart and accessories in a box
    < 0,1 • 0,1 • 0,2 • 0,3 • 0,4 • 0,6 • 0,9 • 1,2 • 2,0 mg/L (Cl₂); 0,10 - 2,00 mg/L (Cl₂)
    DPD
    PF-12Plus; PF-3 Fish; PF-3 Pool; PF-3 Drinking Water; Advance; Visual
    170×125×45 mm (Including packaging)
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 1 830,00 Kč
    1 830,00 Kč
    1 * 1 KIT
       1 830,00 Kč
     : 931016
     : MANA931016
  • VISOCOLOR® ECO Chlorine 2, test no. 5-15; Measuring range on PF-12Plus. Range on other photometers can be different
    VISOCOLOR® ECO Chlorine 2, test no. 5-15; Measuring range on PF-12Plus. Range on other photometers can be different
    MANA931015
    931015
    Reagents for 150 tests, colour chart and accessories in a box
    < 0,1 • 0,1 • 0,2 • 0,3 • 0,4 • 0,6 • 0,9 • 1,2 • 2,0 mg/L (Cl₂); 0,05 - 2,00 mg/L (Cl₂); 0,05 - 2,00 mg/L (Cl₂)
    DPD
    PF-12Plus; PF-3 Drinking Water; Advance; PF-3 Pool; Visual
    170×125×45 mm (Including packaging)
    1 * 1 KIT
    1 * 1 KIT - 1 780,00 Kč
    1 780,00 Kč
    1 * 1 KIT
       1 780,00 Kč
     : 931015
     : MANA931015
  • VISOCOLOR® HE Chlorine
    VISOCOLOR® HE Chlorine
    MANA920015
    920015
    Reagents for 160 tests, colour chart, plastic beaker, 2 round glass tubes, comparator block and one measuring spoon
    0,0 • 0,02 • 0,04 • 0,06 • 0,10 • 0,15 • 0,20 • 0,30 • 0,40 • 0,60 mg/L (Cl₂)
    DPD
    Visual
    240×135×47 mm (Including packaging)
    1 * 1 KIT
    1 * 1 KIT - 3 830,00 Kč
    3 830,00 Kč
    1 * 1 KIT
       3 830,00 Kč
     : 920015
     : MANA920015
  • VISOCOLOR® alpha Chlorine
    VISOCOLOR® alpha Chlorine
    MANA935019
    935019
    Reagents for 150 tests and colour chart in a blister
    0,25 • 0,5 • 1,0 • 1,5 • 2,0 mg/L (Cl₂)
    DPD
    Visual
    154×91×35 mm (Including packaging)
    1 * 1 KIT
    1 * 1 KIT - 827,00 Kč
    827,00 Kč
    1 * 1 KIT
       827,00 Kč
     : 935019
     : MANA935019
  • VISOCOLOR® ECO Schwimming pool
    VISOCOLOR® ECO Schwimming pool
    MANA931090
    931090
    Reagents for 150 tests, colour chart and accessories in a box
    6,9 • 7,2 • 7,4 • 7,6 • 7,8 • 8,2; < 0,1 • 0,1 • 0,2 • 0,3 • 0,4 • 0,6 • 0,9 • 1,2 • 2,0 mg/L (Cl₂)
    Mixed indicator;Chlorine
    Visual
    168×112×40 mm (Including packaging)
    1 * 1 KIT
    1 * 1 KIT - 2 390,00 Kč
    2 390,00 Kč
    1 * 1 KIT
       2 390,00 Kč
     : 931090
     : MANA931090
  • VISOCOLOR® Powder pillows free chlorine, test no. 7-20; *HACH is a registered trademark of HACH company, Loveland, Colorado. The VISOCOLOR product is neither endorsed, nor sponsored by or affiliated with the HACH Company
    VISOCOLOR® Powder pillows free chlorine, test no. 7-20; *HACH is a registered trademark of HACH company, Loveland, Colorado. The VISOCOLOR product is neither endorsed, nor sponsored by or affiliated with the HACH Company
    MANA936220.1
    936220.1
    1000 Powder pillows in a plasitc box
    0,03 - 6,00 mg/L (Cl₂)
    DPD
    UV/VIS II; VIS II; PF-3 Fish; PF-12Plus; PF-3 Drinking Water; PF-3 Pool; Advance
    200×200×260 mm (Including packaging)
    1 * 1.000 KS
    1 * 1.000 KS - 6 310,00 Kč
    6 310,00 Kč
    1 * 1.000 KS
       6 310,00 Kč
     : 936220.1
     : MANA936220.1
Refill packs
  •  
    Contents
    Range
    Chemical method
    For unit
    Dimensions
  • VISOCOLOR® ECO Chlorine 6, refill pack, test no. 5-19;Measuring range on PF-12Plus. Range on other photometers can be different. Only for photometric evaluation.
    VISOCOLOR® ECO Chlorine 6, refill pack, test no. 5-19;Measuring range on PF-12Plus. Range on other photometers can be different. Only for photometric evaluation.
    MANA931219
    931219
    Reagents for 400 tests in a box
    0,05 - 6,00 mg/L (Cl₂)
    DPD
    PF-12Plus; PF-3 Drinking Water; Advance; PF-3 Pool
    170×115×70 mm (Including packaging)
    1 * 1 KS
    1 * 1 KS - 1 750,00 Kč
    1 750,00 Kč
    1 * 1 KS
       1 750,00 Kč
     : 931219
     : MANA931219
  • VISOCOLOR® HE Chlorine, refill pack
    VISOCOLOR® HE Chlorine, refill pack
    MANA920115
    920115
    Reagents for 160 tests in a box
    0,0 • 0,02 • 0,04 • 0,06 • 0,10 • 0,15 • 0,20 • 0,30 • 0,40 • 0,60 mg/L (Cl₂)
    DPD
    Visual
    170×115×70 mm (Including packaging)
    1 * 1 KIT
    1 * 1 KIT - 2 380,00 Kč
    2 380,00 Kč
    1 * 1 KIT
       2 380,00 Kč
     : 920115
     : MANA920115
  • VISOCOLOR® ECO Chlorine 6, refill pack, test no. 5-15; Measuring range on PF-12Plus. Range on other photometers can be different
    VISOCOLOR® ECO Chlorine 6, refill pack, test no. 5-15; Measuring range on PF-12Plus. Range on other photometers can be different
    MANA931215
    931215
    Reagents for 150 tests in a box
    < 0,1 • 0,1 • 0,2 • 0,3 • 0,4 • 0,6 • 0,9 • 1,2 • 2,0 mg/L (Cl₂); 0,05 - 2,00 mg/L (Cl₂); 0,05 - 2,00 mg/L (Cl₂)
    DPD
    PF-12Plus; PF-3 Drinking Water; Advance; PF-3 Pool; Visual
    170×115×70 mm (Including packaging)
    1 * 1 KIT
    1 * 1 KIT - 1 080,00 Kč
    1 080,00 Kč
    1 * 1 KIT
       1 080,00 Kč
     : 931215
     : MANA931215
  • VISOCOLOR® ECO Chlorine 1, refill pack, test no. 5-35; Measuring range on PF-12Plus. Range on other photometers can be different
    VISOCOLOR® ECO Chlorine 1, refill pack, test no. 5-35; Measuring range on PF-12Plus. Range on other photometers can be different
    MANA931235
    931235
    Reagents for 150 tests in a box
    < 0,1 • 0,1 • 0,2 • 0,3 • 0,4 • 0,6 • 0,9 • 1,2 • 2,0 mg/L (Cl₂); 0,05 - 2,00 mg/L (Cl₂); 0,05 - 2,00 mg/L (Cl₂)
    DPD
    PF-12Plus; PF-3 Drinking Water; Advance; PF-3 Pool; Visual
    170×115×70 mm (Including packaging)
    1 * 1 KS
    1 * 1 KS - 1 080,00 Kč
    1 080,00 Kč
    1 * 1 KS
       1 080,00 Kč
     : 931235
     : MANA931235
  • VISOCOLOR® ECO Schwimming pool, refill pack
    VISOCOLOR® ECO Schwimming pool, refill pack
    MANA931290
    931290
    Reagents for 150 tests in a box
    6,9 • 7,2 • 7,4 • 7,6 • 7,8 • 8,2; < 0,1 • 0,1 • 0,2 • 0,3 • 0,4 • 0,6 • 0,9 • 1,2 • 2,0 mg/L (Cl₂)
    Mixed indicator;Chlorine
    Visual
    165×108×47 mm (Including packaging)
    1 * 1 KIT
    1 * 1 KIT - 1 340,00 Kč
    1 340,00 Kč
    1 * 1 KIT
       1 340,00 Kč
     : 931290
     : MANA931290
  • VISOCOLOR® ECO Chlorine 2, refill pack, test no. 5-16; Measuring range on PF-12Plus. Range on other photometers can be different
    VISOCOLOR® ECO Chlorine 2, refill pack, test no. 5-16; Measuring range on PF-12Plus. Range on other photometers can be different
    MANA931216
    931216
    Reagents for 150 tests in a box
    < 0,1 • 0,1 • 0,2 • 0,3 • 0,4 • 0,6 • 0,9 • 1,2 • 2,0 mg/L (Cl₂); 0,10 - 2,00 mg/L (Cl₂)
    DPD
    PF-12Plus; PF-3 Drinking Water; Advance; PF-3 Pool; Visual
    170×115×70 mm (Including packaging)
    1 * 1 KIT
    1 * 1 KIT - 1 240,00 Kč
    1 240,00 Kč
    1 * 1 KIT
       1 240,00 Kč
     : 931216
     : MANA931216
  • VISOCOLOR® ECO Chlorine 6, refill pack, test no. 5-17;Measuring range on PF-12Plus. Range on other photometers can be different. Only for photometric evaluation
    VISOCOLOR® ECO Chlorine 6, refill pack, test no. 5-17;Measuring range on PF-12Plus. Range on other photometers can be different. Only for photometric evaluation
    MANA931217
    931217
    Reagents for 200 tests in a box
    0,05 - 6,00 mg/L (Cl₂); 0,05 - 6,00 mg/L (Cl₂)
    DPD
    PF-12Plus; PF-3 Fish; PF-3 Pool; PF-3 Drinking Water; Advance
    170×115×70 mm (Including packaging)
    1 * 1 KIT
    1 * 1 KIT - 1 280,00 Kč
    1 280,00 Kč
    1 * 1 KIT
       1 280,00 Kč
     : 931217
     : MANA931217