Tisk…
ESD safety shoes, Velcro® fastening, uvex 2 xenova® S1 Trainer, 9504
Dodavatel: Uvex
uvex 2 xenova®
UVEX9504.9/39EA 2450 CZK
UVEX9504.9/39 UVEX9504.8/49 UVEX9504.8/47 UVEX9504.7/38 UVEX9504.9/38 UVEX9504.8/48 UVEX9504.8/45 UVEX9504.8/46 UVEX9504.8/43 UVEX9504.8/44 UVEX9504.7/39 UVEX9504.8/41 UVEX9504.7/50 UVEX9504.8/42 UVEX9504.7/52 UVEX9504.9/51 UVEX9504.8/40 UVEX9504.7/51 UVEX9504.9/52 UVEX9504.9/50 UVEX9504.8/38 UVEX9504.7/47 UVEX9504.8/39 UVEX9504.7/46 UVEX9504.9/48 UVEX9504.7/49 UVEX9504.9/49 UVEX9504.7/48 UVEX9504.7/43 UVEX9504.9/46 UVEX9504.9/47 UVEX9504.7/42 UVEX9504.7/45 UVEX9504.9/44 UVEX9504.7/44 UVEX9504.9/45 UVEX9504.9/42 UVEX9504.8/52 UVEX9504.9/43 UVEX9504.9/40 UVEX9504.8/50 UVEX9504.7/41 UVEX9504.7/40 UVEX9504.8/51 UVEX9504.9/41
ESD safety shoes, Velcro® fastening, uvex 2 xenova® S1 Trainer, 9504
Obuv
Metal-free, lightweight, improved fit and optimum cushioning, padding significantly increase comfort, made of micro velour upper, soft padded collar and outsole made from dual-density PUR sole with excellent slip resistance. Sole materials are free from silicones, plasticisers and other paint-wetting impairment substances.
  • Climazone technology for improved comfort
  • Double individually adjustable velcro
  • Breathable textile lining
  • Removable anti-static comfort insole with moisture control system; with a moisture-wicking system
  • Anti-slip sole with shock absorption at heel and front of the foot
  • 100% metal-free xenova® toecap, compact design, anatomical shape, good lateral stability, no thermal conductivity
  • Fulfils ESD requirements: resistance to ground inferior to 35 megaohms
  • Suitable for indoor and outdoor works
Contains uvex xenova® plastic cap which is the safety feature of this shoes. Suitable for people allergic to chrome, made from synthetic materials.
Colour: Black/red
Certifikace: EN 61340 (ESD); EN ISO 20345:2011; S1, SRC