Tisk…
Agarase
Dodavatel: BioVision
BIOV9201-500EA 7000 CZK
BIOV9201-500
Agarase
A recombinant agarose cleaving protein.
Agarase cleaves agarose to neoagaro-oligosaccharides, and is used for quantitative, gentle recovery of DNA from low melting point agarose.
Upozornení: For research use only. Not to be used in humans.