Cleanroom floor mats, floating systems, CleanZone
Supplier: DYCEM
Certificates
About this item
These floating floor mats are high performance, antimicrobial loose-laid mats offering many benefits without the need for permanent installation. A floating floor can be placed over smooth or rough subfloors and can be self-installed with ease.
- Reduced floor (up to 99,9%) and air particulate (up to 75%) counts in controlled or critical areas
- Long lasting, cost effective solution compared with peel-off mats and other alternatives
- Reduced microbial counts due to Biomaster, a high performance anti-microbial additive
- Easy to clean, maintain, and incorporate into existing Standard Operating Procedures (SOP’s)
- Protected by a standard 12-month warranty from date of installation
Mats are pre-assembled, adhered onto a 1,58 mm thick isolator membrane and cold welded edging to the Dycem material, ensuring a strong bond during its life-cycle. The underside of the floating is an impervious vinyl layer resistant to moisture, which is non-absorbent and contains multiple peduncles in order to minimise sub-floor show through and maintain rigidity.
Certifications: EN 14041:2004
Ordering information: Available colours are cobalt, titanium, midnight, slate, green, or red; contact your Avantor representative for more information.
Product Information
- Catalog #Supplier #L×WThicknessPkAvailabilityPrice
- Floating system floor matFloating system floor matDYCECZ01FS1206CZ01FS12061200×600 mm3,2 mm1Log in for availability1 * 1 KS71 080,00 KčSupplier Article Number: CZ01FS1206Local Article Number: DYCECZ01FS1206Specifications:
Specifications
Environmentally Preferable - Floating system floor matFloating system floor matDYCECZ01FS1206MNCZ01FS1206MN1200×600 mm3,2 mm1Log in for availability1 * 1 KS90 390,00 KčSupplier Article Number: CZ01FS1206MNLocal Article Number: DYCECZ01FS1206MNSpecifications:
Specifications
Environmentally Preferable - Floating system floor matFloating system floor matDYCECZ01FS1210CZ01FS12101200×1000 mm3,2 mm1Log in for availability1 * 1 KS101 720,00 KčSupplier Article Number: CZ01FS1210Local Article Number: DYCECZ01FS1210Specifications:
Specifications
Environmentally Preferable - Floating system floor matFloating system floor matDYCECZ01FS1220CZ01FS12201200×2000 mm3,2 mm1Log in for availability1 * 1 KS192 100,00 KčSupplier Article Number: CZ01FS1220Local Article Number: DYCECZ01FS1220Specifications:
Specifications
Environmentally Preferable - Floating system floor matFloating system floor matDYCECZ01FS1225MCZ01FS1225M1200×2500 mm3,2 mm1Log in for availability1 * 1 KS282 790,00 KčSupplier Article Number: CZ01FS1225MLocal Article Number: DYCECZ01FS1225MSpecifications:
Specifications
Environmentally Preferable - Floating system floor matFloating system floor matDYCECZ01FS2350CZ01FS23501200×4000 mm3,2 mm1Log in for availability1 * 1 KS295 660,00 KčSupplier Article Number: CZ01FS2350Local Article Number: DYCECZ01FS2350Specifications:
Specifications
Environmentally Preferable - Floating system floor matFloating system floor matDYCECZ01FS2006CZ01FS20062000×600 mm3,2 mm1Log in for availability1 * 1 KS87 020,00 KčSupplier Article Number: CZ01FS2006Local Article Number: DYCECZ01FS2006Specifications:
Specifications
Environmentally Preferable - Floating system floor matFloating system floor matDYCECZ01FS2010CZ01FS20102000×1000 mm3,2 mm1Log in for availability1 * 1 KS165 750,00 KčSupplier Article Number: CZ01FS2010Local Article Number: DYCECZ01FS2010Specifications:
Specifications
Environmentally Preferable - Floating system floor matFloating system floor matDYCECZ01FS2010MNCZ01FS2010MN2000×1000 mm3,2 mm1Log in for availability1 * 1 KS249 700,00 KčSupplier Article Number: CZ01FS2010MNLocal Article Number: DYCECZ01FS2010MNSpecifications:
Specifications
Environmentally Preferable - Floating system floor matFloating system floor matDYCECZ01FS2020CZ01FS20202000×2000 mm3,2 mm1Log in for availability1 * 1 KS355 710,00 KčSupplier Article Number: CZ01FS2020Local Article Number: DYCECZ01FS2020Specifications:
Specifications
Environmentally Preferable - Floating system floor matFloating system floor matDYCECZ01FS2030CZ01FS20302000×3000 mm3,2 mm1Log in for availability1 * 1 KS321 090,00 KčSupplier Article Number: CZ01FS2030Local Article Number: DYCECZ01FS2030Specifications:
Specifications
Environmentally Preferable - Floating system floor matFloating system floor matDYCECZ01FS2040CZ01FS20402000×4000 mm3,2 mm1Log in for availability1 * 1 KS430 460,00 KčSupplier Article Number: CZ01FS2040Local Article Number: DYCECZ01FS2040Specifications: