Tisk…
Avantor® ACE® NH₂, HPLC Columns, 3 µm
Dodavatel: VWR
ACE®
ACE111415005SEA 25550 CZK
ACE111415005S ACE111412005S ACE111410005S ACE-1114-75005 ACE-1114-03005 ACE111475005S ACE-1114-15005 ACE-1114-05005 ACE111403005S ACE-1114-12005 ACE-1114-10005 ACE111405005S
Avantor® ACE® NH₂, HPLC Columns, 3 µm
Kolony chromatografické
These stainless steel columns with proprietary bonding technology provide high robustness with excellent column lifetimes.
  • Usable in reversed phase, normal phase, weak anion-exchange and HILIC modes
  • Ideal for the analysis of sugars
  • Ultra-inert, high efficiency with optimum peak shape
  • Range of particle sizes: 1,7 µm, 3 µm and 5 µm and complementary phases