Tisk…
Hypersil™ Silica, HPLC Columns
Dodavatel: VWR
Hypersil™
H5-250AEA 0 CZK
H5-250A HICHH5-10CP3
Hypersil™ Silica, HPLC Columns
Kolony chromatografické
The Hypersil™ classic phases are a global standard for many existing methods. Hypersil is a traditional 120 Å pore size silica.
  • Spherical porous silica
The octyl-, cyano- and phenyl-bonded phases are available as both endcapped and non-endcapped materials.