Tisk…
HPLC columns, Hypersil® MOS C8
Dodavatel: Hichrom Limited
HICHH3MOS-150AEA 14190 CZK
HICHH3MOS-150A HICHH3MOS-50A HICHH3MOS-100A
HPLC columns, Hypersil® MOS C8
The Hypersil® classic phases are a global standard for many existing methods. Hypersil is a traditional 120 Å pore size silica.
  • Spherical porous silica
The octyl-, cyano- and phenyl-bonded phases are available as both endcapped and non-endcapped materials.