Tisk…
Ultra high temperature seal ND20 with central hole, VWR®
VWRI548-2378EA 28910 CZK
VWRI548-2378
Ultra high temperature seal ND20 with central hole, VWR®
High temperature, high quality Silicone/PTFE septum for less extractables at low to mid-high temperatures.
  • Operation upto 300 °C possible.
  • Low bleeding level at high temperatures less than 120 °C