Tisk…

(+)-Sparteine ≥98.0% (by GC), Supelco®

Dodavatel: Merck
Varování

92052-1GEA 5280 CZK
92052-1G
(+)-Sparteine ≥98.0% (by GC), Supelco®
(+)-Sparteine
Vzorec: C₁₅H₂₆N₂
MW: 234,39 g/mol
MDL Number: MFCD00869353
CAS číslo: 492-08-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR