1,8-Anthracenedimethanamine ≥98.0% (by HPLC), Supelco®
Dodavatel: Merck
|
Varování
|
11353-50MGEA
0
CZK
11353-50MG
1,8-Anthracenedimethanamine ≥98.0% (by HPLC), Supelco®
1,8-Anthracenedimethanamine
Vzorec:
C₁₆H₁₆N₂ |
CAS číslo:
853785-51-0 |
Výsledky specifikačního testu
Appearance (Color) | Faint Grey to Grey and Faint Brown to Brown |
Appearance (Form) | Powder or Crystals |
Purity (HPLC area %) | ≥ 98.0 % |
Carbon Content | 81.32 % (Theory) |
Hydrogen Content | 6.82 % (Theory) |
Nitrogen Content | 11.85 % (Theory) |
Proton NMR Spectrum | Conforms to structure |
RECOMMENDED RETEST PERIOD | 36 MONTHS |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...